Hyaluronidase
Lieferant: STEMCELL TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
07462 07461
STMC07462EA 671 EUR InStock
STMC07462 STMC07461
Hyaluronidase
Zellkultur-Ergänzungen und -Zusätze
For hydrolysis of hyaluronic acid.
Hydrolize hyaluronic acids in connective tissue with Hyaluronidase. Frequently used with other proteases, such as collagenase, this polysaccharidase cleaves endo-N-acetlyhexosaminic bonds in hyaluronic acid, as well as chondroitin sulfate A and C, to tetrasaccharide residues. Hyaluronidase consists of a single polypeptide chain of 450 amino acid residues with four disulfide bonds and is glycosylated, containing 5% mannose and 2.17% glucosamine (Borders and Raftery).
Achtung! Bitte beachten: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge