CoV Protease Substrate-2 IF670
Lieferant: STEMCELL TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100-0512
STMC100-0512EA 0 EUR InStock
STMC100-0512
CoV Protease Substrate-2 IF670
Proteine und Peptide
FRET-based peptide substrate for detection of protease activity of coronaviruses - 1000 tests.
  • Enables efficient detection of coronavirus protease activity
  • Can be used as a high throughput screening tool for inhibitors of coronavirus proteases
CoV Protease Substrate-2 IF670 allows for efficient detection of coronavirus (CoV) proteases. This fluorescence resonance energy transfer (FRET)-based peptide substrate can be cleaved by 3C-like proteases (3CLpro), the CoV main protease involved in regulating the coronavirus replication complex. 3CLpro is also a potential target for therapeutic applications.
CoV Protease Substrate-2 IF670 has an excitation wavelength of 656 nm, an emission wavelength of 670 nm, an extinction coefficient of 250000 cm⁻¹ M⁻¹. It consists of a 12 amino acid sequence (VNSTLQSGLRKM) that also contains the Tide Quencher™ 5 (TQ5; quencher) and iFluor™ 670 (donor) fragments at the N- and C- termini, respectively. When the peptide substrate is intact, favorable energetic overlap of the iFluor™ 670 excited state and the TQ5 absorption leads to efficient intramolecular fluorescence quenching of iFluor™ 670 by the TQ5 fragment. Cleavage of the substrate by CoV proteases, however, dissociates the iFluor™ 670 fragment from TQ5, restoring the fluorescence of the iFluor™ 670 moiety that is directly correlated to the CoV protease activity.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge