SARS-CoV-2 Recombinant Spike Protein, aa16-685 (HEK293-expressed), FLAG and His tags
Lieferant: STEMCELL TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100-0594 100-0595
STMC100-0594EA 1670 EUR InStock
STMC100-0594 STMC100-0595
SARS-CoV-2 Recombinant Spike Protein, aa16-685 (HEK293-expressed), FLAG and His tags
Proteine und Peptide
SARS-CoV-2 Recombinant Spike Protein, aa16-685 is expressed in HEK293 cells and is one of four structural proteins encoded by the SARS-CoV-2 genome. The Spike Protein plays a key role in attachment to host cells, allowing invasion through clathrin-mediated endocytosis. The Spike Protein can be cleaved by host cell proteases after aa685 to yield the N-terminal S1 subunit and C-terminal S2 region. The S1 subunit is responsible for interacting with the host cell receptor (angiotensin-converting enzyme II) through a receptor-binding domain that is highly conserved with SARS-CoV. The S1 subunit has two conformations: a ‘down’ conformation in which the receptor is inaccessible, and an ‘up’ conformation in which the receptor is accessible. These conformational changes are key for monoclonal antibody drugs and vaccine development. SARS-CoV-2 Recombinant Spike Protein contains a polyhistidine tag at the amino terminus; it also contains a FLAG tag at the carboxy terminus.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge