SARS-CoV-2 (ORF3a) Peptide Pool
Lieferant: STEMCELL TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
100-0649
STMC100-0649EA 0 EUR InStock
STMC100-0649
SARS-CoV-2 (ORF3a) Peptide Pool
Proteine und Peptide
The SARS-CoV-2 (ORF3a) Peptide pool is a lyophilised mixture of 66 peptides from open reading frame 3a (ORF3a) protein of SARS-CoV-2. ORF3a is found in both SARS-CoV and SARS-CoV-2 and the two proteins share high sequence similarity. SARS-CoV ORF3a has been shown to cause apoptosis of infected cells and form potassium-sensitive ion channels, modulating the release of viruses. Additionally, it induces ligand-independent downregulation of Type 1 interferon receptor. The sequence similarity between the two proteins suggests it may play a similar role in SARS-CoV-2. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 275 on SARS-CoV-2 ORF3a.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge