Human Recombinant NT-4
Lieferant: STEMCELL TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
78093 78093.1
STMC78093EA 282 EUR InStock
STMC78093 STMC78093.1
Human Recombinant NT-4
Proteine und Peptide
Neurotrophin-4 (NT-4) is a member of the nerve growth factor family which includes neurotrophin-3 (NT-3), brain-derived neurotrophic factor (BDNF), and nerve growth factor (NGF), all of which promote the differentiation, growth, and survival of peripheral and central nervous system neurons (Eide et al.). NT-4 binds and activates tropomyosin receptor kinase B (TrkB) at the cell surface; in doing so, it acts as a survival factor for certain populations of sensory neurons (Berkemeier et al.; Skaper). It has been shown that NT-4, together with BDNF, promotes neurite extension and maturation, as well as maintenance of differentiated cerebellar granule cells (Gao et al.).
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge