Human Recombinant GRO-beta (CXCL2)
Lieferant: STEMCELL TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
78112.1 78112.2
STMC78112.1EA 349 EUR InStock
STMC78112.1 STMC78112.2
Human Recombinant GRO-beta (CXCL2)
Proteine und Peptide
GRO (growth-regulated oncogene)-beta or CXCL2 is a member of the CXC family, which plays an integral role in recruitment and activation of neutrophils and basophils in response to tissue injury and microbial infection. GRO-beta and GRO-gamma are closely related to GRO-alpha and share 90% and 86% amino acid sequence homology, respectively, with GRO-alpha. Receptor-binding studies have demonstrated that GRO-alpha, -beta, and -gamma signal mainly through G protein-coupled receptors CXCR1 and CXCR2 (Ahuja and Murphy). GRO-beta is expressed in epithelial cells, monocytes, fibroblasts, and melanocytes and is further induced during inflammatory, epithelialization, and angiogenic processes, for example during the wound healing process of human burn wounds (Zaja-Milatovic and Richmond). GRO-beta also stimulates mitogenesis in certain human melanoma cells (Unemori et al.).
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge