Lambda ZAP II vector kits
Lieferant: STRATAGENE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
236612
STRA236612EA 2360 EUR InStock
STRA236612
Lambda ZAP II vector kits
Vektoren, Plasmide und Bibliotheken
The Lambda ZAP II vectors combine the high efficiency of lambda vector systems with the versatility of plasmid systems.
They enable prokaryotic expression of your gene of interest under control of the lac promoter.
in vivo excision of the Lambda ZAP II vector yields the pBluescript SK- vector, which is a high-copy number pUC-based phagemid.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge