Stainless steel bubblers, vertical, electropolished with fill-port
Lieferant: Strem Chemicals, Inc.
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
96-4998.1
STRE96-4998.1EA 0 EUR InStock
STRE96-4998.1
Stainless steel bubblers, vertical, electropolished with fill-port
Blasenzähler
Chemical vapor deposition/atomic layer deposition (CVD/ALD) equipment.
  • PCTFE valve stem tip (121 °C)
  • Standard temp. valve
Volatile precursors for CVD.
Bestellinformation: For dimensions or more information, contact your Avantor representative.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge