Bovine native alpha-A-Crystallin
Lieferant: STRESSGEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SPP-226F
STSSSPP-226FEA 0 EUR InStock
STSSSPP-226F
Bovine native alpha-A-Crystallin
Proteine und Peptide
Alpha A crystallins (known as CRYAA, HspB4, heat shock protein β-4) are major water-soluble proteins accounting for almost 50% of total protein in the mammalian transparent eye lens and they are also found in a variety of other tissues.
  • Isolated from bovine eye lens
  • Purified by multi-step chromatography (SDS-PAGE, western blot)
  • Recommended dilutions (conditions) include western blot (100 ng/lane, colorimetric)
It has a liquid formulation in PBS at pH 7,2 containing 0,15M sodium chloride and 0,05M phosphate buffer.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge