Human recombinant HSP27 (from E. coli)
Lieferant: STRESSGEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SPP-715F
STSSSPP-715FEA 0 EUR InStock
STSSSPP-715F
Human recombinant HSP27 (from E. coli)
Proteine und Peptide
Human Hsp27, mouse Hsp25 and ab-crystallin are part of a diverse family of small heat shock proteins which are produced in all organisms. They function as chaperone-like proteins by binding unfolded polypeptides and preventing uncontrolled protein aggregation.
  • Produced in E. coli
  • Purified by multi-step chromatography (SDS-PAGE, western blot)
  • Recommended dilutions (conditions) include western blot (100 ng, colorimetric)
It has a liquid formulation in 20 mM TRIS at pH 7,5, containing 10 mM sodium chloride, 1 mM EDTA, and 1 mM DTT.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge