Bovine Recombinant HSC70/HSP73 (from E. coli), Biotin
Lieferant: STRESSGEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SPP-761E
STSSSPP-761EEA 0 EUR InStock
STSSSPP-761E
Bovine Recombinant HSC70/HSP73 (from E. coli), Biotin
Proteine und Peptide
The Hsp70 family of heat shock proteins contains multiple homologs ranging in size from 66 to 78 kDa, and are the eukaryotic equivalents of the bacterial DnaK. Hsp70 nucleotide exchange and substrate binding coordinates the folding of newly synthesised proteins, the re-folding of misfolded or denatured proteins, coordinates trafficking of proteins across cellular membranes, inhibits protein aggregation, and targets the degradation of proteins via the proteasomal pathway.
  • Produced in E. coli
  • Purified by multi-step chromatography (SDS-PAGE, western blot)
It has a liquid formulation in 10 mM sodium phosphate at pH 7,2.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge