Cole-Parmer® HP-200 Analogue Hotplate with PTFE Platform
Lieferant: Cole Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
04801-08
STUA04801-08EA 5150 EUR InStock
STUA04801-08
Cole-Parmer® HP-200 Analogue Hotplate with PTFE Platform
Heizgeräte Heizplatten
Impervious to chemical attack, even concentrated acids.
  • Specifically designed for acid digestions and trace metal analysis
  • Separate temperature controller allows safe placement outside the fume hood
  • Large, square top plate accommodates one large vessel or several smaller ones
  • Plate is rated IP 43; controller is rated IP 30
With a glass ceramic heating plate mounted in a solid block of pure PTFE, this rugged hotplate withstands boiling acid solutions and aggressive cleaning agents. The separate analogue temperature controller is connected via a 2 m PTFE coated cable, allowing it to be placed away from corrosive environments. Hot warning light flashes when temperature is above 122 °F (50 °C).
Lieferumfang: Supplied with power cord with Schuko and UK plugs.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge