Cole-Parmer® RC-200 Recirculating Cooler
Lieferant: Cole Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
12152-77
STUA12152-77EA 5030 EUR InStock
STUA12152-77
Cole-Parmer® RC-200 Recirculating Cooler
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
Powerful cooling with a compact footprint.
  • Optimise accuracy with easy temperature offset calibration
  • Worry free operation - visual safety alarm indicates overload
  • Easy to read current temperature readout on LED display
This reliable, easy to use recirculating cooler offers accurate and powerful cooling for an external device. Cooler can achieve temperatures down to −20 °C, making it ideal for controlling equipment in temperature-sensitive applications.
Featuring convenient temperature regulation, the LED display features the current temperature, alarm indicator, and press button temperature set point control. An easy-to-use calibration function allows user to adjust the temperature display to match a traceable temperature standard.
Use the 9 mm ID hose barb inlet/outlet ports to connect to a variety of external equipment. A dedicated drain port allows easy emptying and cleaning.
Lieferumfang: Supplied with bath cover, two spare fuses and power cord with UK 3-pin and Schuko 2-plug plugs.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge