Cole-Parmer® SH-200 Reciprocating Shakers
Lieferant: Cole Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
51901-02
STUA51901-02EA 2490 EUR InStock
STUA51901-02
Cole-Parmer® SH-200 Reciprocating Shakers
Schüttler Laborschüttler, reziprok
Powerful yet quiet device is perfect for continuous use applications.
  • Digital speed selection provides consistent mixing
  • Soft-start motor prevents spills and splashes
  • Fully adjustable cradle accommodates most flasks, bottles, and beakers
  • Can be used in incubator systems
These linear reciprocating shakers feature digital speed selection from 25 to 250 min⁻¹ with a soft start. Four adjustable rubber cushioned horizontal securing bars with quick release handles on the cradle-type platform allow users to fit nearly any size flask, bottle, or beaker. Secure up to twelve 250 ml, nine 500 ml, four 1 L, or two 2 L vessels.
Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Shaker is capable of continuous use, and the microprocessor maintains speed accuracy over long runs.
Lieferumfang: Unit includes fully adjustable cradle system for flasks and bottles and power cord.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge