Cole-Parmer® GP Laboratory Overhead Mixer
Lieferant: Cole Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
55008-00
STUA55008-00EA 0 EUR InStock
STUA55008-00
Cole-Parmer® GP Laboratory Overhead Mixer
Rührer Rührwerke
Mixer designed for reliability and durability.
  • Ideal for light-duty mixing of water-like fluids
  • Brushed DC motor with integrated overload protection
  • Durable - no wearing parts that require user service
  • User-replaceable keyless chuck
  • 1/50 hp brushed DC motor
The Cole-Parmer GP laboratory mixer ensures accurate, dependable motor speed control with 'soft-start' feature and smooth, gradual adjustment to the speed setting. Constant speed is maintained electronically via automatic compensation for changes in load torque demands at both high and low speeds. Built-in motor current compensation minimises speed drift with changes in load. Motor current limiting protects the system in case of a locked rotor condition. Output circuit protection protects against inadvertent output shorting.
The mixer rotates at 40 to 4000 min⁻¹ with a maximum torque of 9,5 in-oz (6,7 N·cm). The speed control knob is used to turn on and set the speed of the mixer motor and provides a 100:1 adjustment range. The LED indicates the mixer is powered on when it is green. When it flashes, it indicates a limit condition has been reached, such as an over temperature or over-current situation.
Shaft: L×Ø: 305×10 mm
Motor: L×H 84×104 mm
Material: Painted or anodised aluminum, plastic, and vinyl
Zertifizierungen: IP protection class according to DIN EN 60529: IP 44.
Lieferumfang: Includes Mixer motor with integrated controller and 183 cm (6') cable, 12× 0.95 cm (³/₈") stainless steel shaft, 380 mm (1,5") stainless steel propeller, mounting rod and power cable.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge