Surgical blade, No. 3, stitch cutter
Lieferant: SWANN MORTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0326
SWAN0326EA 82.1 EUR InStock
SWAN0326
Surgical blade, No. 3, stitch cutter
Skalpelle Skalpellklingen
The stainless steel No. 3 or 'baby' stitch cutter is used in conjunction with the No. 3 surgical handle.
  • Convenient and cost effective
  • Sterile
Enables a more aseptic techinique by operating further away from the actual wound which also allows better visibility.
Fits handles 3 Graduated, 3L, 5B, 7, 9, B3 and B3L.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge