Schneidwerkzeug
Lieferant: SWANN MORTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1231
SWAN1231EA 68.7 EUR InStock
SWAN1231
Schneidwerkzeug
Skalpelle Skalpelle, komplett
Klinge aus Stahl, mit Kunststoff-Handgriff. Das ideale Werkzeug für viele Schneid- und Modellierarbeiten.
  • Thumbscrew is used for blade changing and retaining
  • Handle made from PS is impact resistant
Three types of replacement blades available as an option
Bestellinformation: This starter set includes the plastic handle and two blades (one each No. 1 and No. 2 (carded)). Handle and blades are also available separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge