Surgical blades for cervical biopsy
Lieferant: SWANN MORTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2001
SWAN2001EA 135 EUR InStock
SWAN2001
Surgical blades for cervical biopsy
Skalpelle Skalpellklingen
This sterile stainless cervical biposy blade is used in conjunction with the long stainless steel Major handle to take a 'cone' biopsy from the tissue in a woman's cervix. The tissue sample can then be analysed to assess the prescence of malignant and non malignant carcinoma cells.
  • Fits handles Major S and L
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge