Myringotomy blade
Lieferant: SWANN MORTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2102
SWAN2102EA 73.1 EUR InStock
SWAN2102
Myringotomy blade
Skalpelle Skalpellklingen
The sterile stainless steel Myringotomy blade is used in conjunction with the SF1 or SF13 Fine surgical handle in the treatment of 'Otitis media' or 'Glue ear'.
  • Blade and handle made of stainless steel
  • Blade is double egded so allowing for cutting movements in both directions
  • Fits handles SF1, SF2, SF3, SF4, SF13 and SF23
The Myringotomy blade is used to make an incision in the tympanic membrane (ear drum) into which a 'grommet' can be placed to allow ventilation of the middle ear which has become blocked thus interfering with the patients ability to hear.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge