Disposable scalpels, Minor
Lieferant: SWANN MORTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2381 2373 2382 2385 2386 2383 2371 2372 2375 2376
SWAN2381EA 20.7 EUR InStock
SWAN2381 SWAN2373 SWAN2382 SWAN2385 SWAN2386 SWAN2383 SWAN2371 SWAN2372 SWAN2375 SWAN2376
Disposable scalpels, Minor
Skalpelle Skalpelle, komplett
These non sterile disposable scalpels consist of a stainless steel blade and a PS handle.
  • 'Thumb' scalpels with a protective guard
  • Ideal for surgical applications
Please note: The protective guard is NOT compatible with a standard metal handle and blade.
Bestellinformation: Available in packets of 10 or bulk boxes of 100.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge