Silvers skin graft knives
Lieferant: SWANN MORTON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9911 9912
SWAN9911EA 1660 EUR InStock
SWAN9911 SWAN9912
Silvers skin graft knives
Skalpelle Skalpelle, komplett
These miniature skin graft handles are to be used with surgical stainless double edged razor blades. Ideal for harvesting small intricate grafts from areas which would not be accessible with the larger standard knives such as the ear, dorsa of the hands or scalp.
  • Non sterile
  • Made from stainless steel
  • Available with solid stainless steel or distinctive blue autoclavable handle
Although used mainly within plastic, reconstructive and oral surgery the graft handles can also be used for the debridement of wounds.
Verpackung: Individually boxed.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge