Western blots, pre-made, mouse subcellular fraction specific blot
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TB47
TB47EA 2030 EUR InStock
TB47
Western blots, pre-made, mouse subcellular fraction specific blot
Elektrophorese-Reagenzien Western Blotting Reagents
The subcellular fractions from liver tissue are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse livers.
Each blot contains the following mouse liver subcellular fractions: Whole liver, nuclei, mitochondria, Golgi complex, smooth endoplasmic reticulum, and rough endoplasmic reticulum.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge