Resolvex® M10 Manual Positive Pressure Processor
Lieferant: TECAN TRADING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
288-0001 288-0006 289-0004
TCAT288-0001EA 0 EUR InStock
TCAT288-0001 TCAT288-0006 TCAT289-0004
Resolvex® M10 Manual Positive Pressure Processor
Verteiler
Benchtop manual positive pressure processor.
  • Individual flow restrictors ensure parallel processing even when some wells are vacant
  • Only a gas supply is needed for operation
  • Dual-level gas delivery system offers fine controls of flow and pressure
Perfect for entry into positive pressure processing and workflows, the intuitive design of the Resolvex® M10 makes it easy to use with minimal training required.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge