Cerex® RCH PPBA 96 Well Plate
Lieferant: TECAN TRADING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30162759
TCAT30162759EA 0 EUR InStock
TCAT30162759
Cerex® RCH PPBA 96 Well Plate
Mikrotiterplatten Festphasenextraktionsplatten
A special phenyl boronic acid chemistry on polymer base resin for better capacity and solvent use.
  • Cerex
  • Reversed-phased
  • Liquid handler compatible
PBA utilises an unique covalent interaction ideal for catecholamine analysis.
Verpackung: 1 piece per box
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge