Cerex® FLORASIL SPE Columns
Lieferant: TECAN TRADING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30213512
TCAT30213512EA 0 EUR InStock
TCAT30213512
Cerex® FLORASIL SPE Columns
Chromatographiesäulen
Florisil is used for sample preparation (solid phase extraction) of e.g., organic tin compounds and aliphatic carboxylic acids. Florisil is a magnesium silicate.
  • Ion-exchange
  • Liquid handler compatible
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge