Cerex® SPE Columns HPWCX
Lieferant: TECAN TRADING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30174772 30215446
TCAT30174772EA 0 EUR InStock
TCAT30174772 TCAT30215446
Cerex® SPE Columns HPWCX
Chromatographiesäulen
Polymer monodisperse weak cation exchange chemistry, used for retention of strong bases in biological fluids. Ideal for strong base extraction for drug monitoring, screening, identification, confirmation, and quantitation.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge