SPE Plates, 96-well, NBE HPSCX
Lieferant: TECAN TRADING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
427-0021NBE 427-0051NBE
TCAT427-0021NBEEA 0 EUR InStock
TCAT427-0021NBE TCAT427-0051NBE
SPE Plates, 96-well, NBE HPSCX
Mikrotiterplatten Festphasenextraktionsplatten
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
  • Ion-exchange
  • Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge