Cerex® TRACE-N20 SPE Column
Lieferant: TECAN TRADING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
611-0101R
TCAT611-0101REA 0 EUR InStock
TCAT611-0101R
Cerex® TRACE-N20 SPE Column
Chromatographiesäulen
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
  • Reversed-phase
  • Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge