Microplate washer, HydroSpeed™
Lieferant: TECAN TRADING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91200001
TCAT91200001EA 0 EUR InStock
TCAT91200001
Microplate washer, HydroSpeed™
Mikrotiterplatten-Washer
The HydroSpeed is Tecan’s advanced plate washer, designed for quick, parallel washing of entire 96- and 384-well plates. This avoids time differences between the wells for uniform results. In addition, the HydroSpeed provides full control of wash power for optimized washing of cells, beads and ELISAs.
  • Cell protection - extra gentle cell washing with tuneable wash power
  • Adjustable wash head positions, speed settings and vacuum level
  • Anti-clogging - active prevention of needle clogging
  • Easy X-change wash heads for intense cleaning in ultrasonic bath
  • Automated buffer switching of four liquids and plate shaking capability
  • Predefined plate library and protocols for an easy start
  • Plate assistant function for easy teaching of new plates
The HydroSpeed plate washer is ready for ELISA washing, as well as gentle washing of adherent and weakly-adherent cells. It supports soak steps with microplate shaking, as well as buffer switching.
With full on-board operation via the easy-to-use touchscreen interface, the HydroSpeed really does offer you power and control for all your cell, bead and ELISA washing applications.
Bestellinformation: Accessories not included (must be ordered seperate). Additional configuration available on request.
Lieferumfang: Includes 1× Washer, 1× 10 L set bottle waste, 2× 4 L wash bottles, 1× vacuum pump, 1× power cable, 1× USB stick with manuals in 11 languages.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge