Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Microplate readers, Infinite® 200 PRO

Lieferant: TECAN TRADING

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

92000001 92100001 92200001 92300001 92400001 92500001
TCAT92000001EA 0 EUR InStock
TCAT92000001 TCAT92100001 TCAT92200001 TCAT92300001 TCAT92400001 TCAT92500001
Microplate readers, Infinite® 200 PRO
Mikrotiterplatten-Photometer
The Infinite 200 PRO plate reader family offers affordable detection solutions empowered by monochromator- or filter based optics.

  • Quad4 Monochromators for free wavelength selection
  • Spectral scanning in both absorbance and fluorescence modes
  • Optimised detectors for each reading mode
  • Filter optics for TR-FRET, HTRF® and FP reading modes
  • z-focussing as integrated background correction
  • Field upgradeable injectors
  • Supports 6 to 384 well plates
  • DNA-, RNA- & protein quantification
  • Low sample volume: 2 µl
  • Up to 16 sample in parallel
  • A260/280 purity check
  • Labeling effiency for nucleic acids
  • Full-spectra scan (Spark, SparkCyto)

The Infinite 200 PRO comes in six tailored instrument configurations for research labs providing excellence in ELISA assays, nucleic acid quantifications, reporter assay technologies, and drug discovery assays, including HTRF® and fluorescence polarisation.
Dimensions (W×D×H): from 425×457×253 mm.

All Infinite 200 microplate readers configurations include:
• Linear and orbital plate shaking
• Temperature control: 5 °C above ambient to 42 °C
• Light source: High energy Xenon flash lamp
• Supported plate formats: 6 to 384 well plates
• USB-connection interface
• i-control™ reader control software

Optional items:
• Injectors for reagent dispensing

NanoQuant Plate allows parallel quantification and analysis of up to 16 nucleic acid or protein samples, in sample volumes as little as 2 μl. For convenience and optimal data quality, the Nano-Quant Plate is the only low volume plate on the market that is 100% calibration free.

Bestellinformation: Accessories not included (must be ordered seperately).

Lieferumfang: Instruments supplied with i-control™ reader control software, user manuals as pdf-files, and power cable.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge