Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® FSB-200 Series Compact Fluidised Sand Baths

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

01189-50 01184-00 01184-02
TECN01189-50EA 7119.8 EUR InStock
TECN01189-50 TECN01184-00 TECN01184-02
Cole-Parmer® FSB-200 Series Compact Fluidised Sand Baths
Bäder Wirbelbett-Bäder
Fast heat-ups and accurate temperature control to 600 °C.

  • Dry, inert, non-toxic aluminium oxide ensures hazard-free operation
  • Safer alternative to high temperature oil and salt baths
  • Requires controller in order to be calibrated
  • Material: Stainless steel

Use these fluidised sand baths for heat treating processes, temperature sensor calibrations and electrical component testing. Baths use aluminium oxide fluidised by low-pressure air as a dry bath medium with excellent heat transfer properties. Inert, non-flammable aluminium oxide is safer to use than oil and will not corrode sample vessels or produce toxic fumes. Vessels of all shapes can be easily immersed and safely removed from the bath, without the hazard and mess of hot oils.

Make temperature adjustments via a control knob and reference dial. Baths require a dry air flow of approximately 57 L/min at about 3 psi pressure; order an air pump or temperature controller to accurately control the temperature of the sand bath separately. Baths feature a thermowell to hold a thermocouple thermometer.

Lieferumfang: Supplied with overspill flange and power cord with plug. Supplied with Type K thermocouple probe and power cord.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge