Cole-Parmer® WBU-200 Series Unheated Water Baths
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
16101-88 16101-89 16104-45 16104-53 16112-01
TECN16101-88EA 596.46 EUR InStock
TECN16101-88 TECN16101-89 TECN16104-45 TECN16104-53 TECN16112-01
Cole-Parmer® WBU-200 Series Unheated Water Baths
Bäder Wasserbäder, ohne Umwälzung
Corrosion resistant baths with stainless steel inners for easy cleaning.
  • Constructed of durable stainless steel
  • Rugged splash-proof case
  • Integrated carrying handle for added safety
  • Material: Stainless steel
Designed to be used with the Cole-Parmer IC-300 series immersion circulators (sold separately), these water baths incorporate carrying handles for added safety. All water baths have stoved enamelled steel outer cases and are supplied with bridge mounting plate to hold the thermoregulator.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge