Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cole-Parmer® WBU-200 Series Liquid Calibration Baths

Lieferant: Cole-Parmer

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

16104-50 16105-04 16112-03
TECN16104-50EA 1863.15 EUR InStock
TECN16104-50 TECN16105-04 TECN16112-03
Cole-Parmer® WBU-200 Series Liquid Calibration Baths
Umwälzthermostate Offene Umwälzsysteme Kalibrier-Thermostate
Compact and accurate water baths for external circulation or temperature calibration.

  • Fully insulated baths offer excellent heat retention
  • Temperature stability of ±0.005 °C depending on choice of control unit

These compact calibration baths offer excellent stability over the entire temperature range. Use these
baths for external circulation to maintain temperatures of samples in viscometers, photometers, refractometers, fermenters, and other reaction vessels.

All models offer high pump performance and exceptional thermal stability from −35 to 250 °C. Add a flow cooler to the system to achieve temperatures down to −35 °C. The water baths are fully insulated on all sides and base and are fitted with a cooling coil for connection to a cold water supply for use at temperatures around ambient.

An immersion circulator is required for liquid calibration baths to operate - order IC-300 series immersion circulators separately.

Lieferumfang: Supplied with lid, drain tap, carry handles, a cooling coil (with bung) and hole to position a certified sensor.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge