Cole-Parmer® CB-200D-IB Electronic Ice Bucket
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
24120-00
TECN24120-00EA 1621.46 EUR InStock
TECN24120-00
Cole-Parmer® CB-200D-IB Electronic Ice Bucket
Blockthermostate Heiz-/Kühl-Blockthermostate
Keep samples cold, clean, and dry.
  • Waterless - no more wet, melting ice or floating sample tubes
  • Ceramic-coated beads maintain a 4 °C temperature for the entire working day
  • Holds approximately 60×1,5 ml microtubes
Use the electronic ice bucket to keep tubes clean and dry while incubating at sub-ambient temperatures for long periods of time. A removable bucket is filled with ceramic-coated beads that can maintain a constant temperature and support the sample tubes. To save time, the bucket of beads can be incubated in the refrigerator overnight and inserted into the unit before use. Reusable beads can be autoclaved at 134 °C (273 °F).
A temperature control ensures all contents in the ice bucket are kept at an even temperature, whether cold or warm. A dark plastic lid helps maintain a constant temperature and protects light sensitive samples. View the current temperature on a five-digit orange LED display in either °C or °F.
Lieferumfang: Supplied with aluminum bucket, bag of ceramic-coated beads, dark plastic lid and power cord.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge