Cole-Parmer® CB-200 Block Chiller/Heater
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36620-17
TECN36620-17EA 1341.96 EUR InStock
TECN36620-17
Cole-Parmer® CB-200 Block Chiller/Heater
Blockthermostate Heiz-/Kühl-Blockthermostate
Safely maintain biological samples.
  • Designed for maintaining biological samples safely between 4 and 37 °C
  • Alarm sounds when set temperature is reached and elapsed time ends
  • Five-digit LED display can be set for either °C or °F
  • Choose from a wide range of interchangeable aluminium block inserts
This benchtop block chiller/heater is ideal for incubating sample tubes at temperatures ranging from 0...40 °C (32...104 °F). The Isotemp® temperature control ensures all contents are kept at an even temperature, whether cold or warm. Integrated countdown/count-up timer with audible alarm allows up to nine days to be programmed and will not start until a stable temperature has been reached.
Use with Dri-Block® aluminium inserts to maintain temperature stability of ±0,2 °C. Unit holds two single-width inserts or one double-width inserts. A dark plastic lid protects light-sensitive reagents as well as helping to maintain the sample temperature.
Lieferumfang: Lid and power cords with plug (US, UK, and EU).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge