Mesh Stainless Steel Locker
Lieferant: TEKNOMEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CRLO-ME24D
TEKMCRLO-ME24DEA 0 EUR InStock
TEKMCRLO-ME24D
Mesh Stainless Steel Locker
Möbel Garderobenschränke
Our stainless steel mesh storage locker with hasps and stables is a ideal storage solution. With 24 independently lockable storage cages incorporating a hygienic design.
  • 304-grade stainless steel
  • Brushed dull polish finish
  • 24 Compartment
  • Fitted with 15 cm high legs
  • All doors fitted with hasp and staple ready for use with padlocks
  • Compartment: 305×305×220 mm
  • Floor Clearance: 150 mm
  • Mesh grid: 25×25 mm
This locker comes in a three by eight configuration making it ideal for ear defenders, personal items or hard hats.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge