Couplings for chest racks
Lieferant: TENAK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TE76220 TE76200
TENATE76220EA 32 EUR InStock
TENATE76220 TENATE76200
Couplings for chest racks
Gestelle Kryogestelle
The coupling for chest racks has ergonomic working position, when operating with racks in a chest freezer, it can be a great help to stack two racks on top of each other.
Racks which are often used can be on top and long term storage can be in the bottom. In order to secure the top rack we have designed this coupling.
It is available for TENAK Classic and Comfort racks for standard boxes and rack for microtiterplates.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge