Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

SpeedVac™ SPD1030P1 Integrated Vacuum Concentrator System Kit

Lieferant: THERMO SAVANT

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

SPD1030P1-230
THSASPD1030P1-230EA 23400 EUR InStock
THSASPD1030P1-230
SpeedVac™ SPD1030P1 Integrated Vacuum Concentrator System Kit
Konzentratoren
Thermo Scientific™ Savant™ SpeedVac™ SPD1030 and SPD2030 vacuum concentrators are fully integrated systems that combine a concentrator, refrigerated cold trap, diaphragm pump and a vacuum gauge into a single, compact unit.

  • Proven reliability: Over 50 years of experience with an oil-free PTFE diaphragm vacuum pump for virtually maintenance-free operation
  • Speed and efficiency: Rapid evapouration and solvent removal with a radiant cover and vacuum controls to prevent sample bumping
  • Ease of use: Intuitive design with a bright display, three preset and three user-defined programs, and an oil-free vacuum source. USB port for data transfer
  • Safety assurance: Soft-close glass lids lock securely for the safety of samples and technicians

The Savant SpeedVac SPD1030 system is designed for medium capacity (i.e., 120×1.5 ml tubes and other formats) while the Savant SpeedVac SPD2030 system accommodates higher capacity (i.e., 200×1.5 ml tubes and other formats). A variety of rotors are available for both systems to support sample formats from micro-centrifuge tubes, vials, and microplates to large-volume flasks. Both systems fit in a standard laboratory fume hood.

Bestellinformation: SPD1030P1-230 includes acid-resistant SpeedVac™ vacuum concentrator, rotor accessory, glass condensation flask, CryoCool™ heat transfer fluid (1 L).

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge