Savant™ SpeedVac™ SPD140P2 Medium Capacity Vacuum Concentrator for Combinatorial Chemistry Applications
Lieferant: THERMO SAVANT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SPD140P2-230
THSASPD140P2-230EA 40000 EUR InStock
THSASPD140P2-230
Savant™ SpeedVac™ SPD140P2 Medium Capacity Vacuum Concentrator for Combinatorial Chemistry Applications
Konzentratoren
Savant™ SpeedVac™ medium capacity vacuum concentrators evapourate strong acids, bases, and aggressive organic solvents. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
  • Proven reliability: Over 50 years of experience with the Savant SpeedVac products
  • Consistent drying: Radiant lamps, PTFE-coated chamber, and heated glass lid ensure efficient drying of various solvents and prevent condensation
  • User-friendly: Intuitive interface with four preset and eight user-defined programs for temperature, ramp rate, and vacuum level settings
  • Safety features: Soft-close, secure-lock lid, hands-free rotor loading, and audible operation completion alarm
  • Data transfer: USB port for live run data download
Solvent and solvent combinations for SPD140P1 kit are acetic acid, ammonium hydroxide, formic acid, hydrochloric acid, TFA, methylene chloride and TFA, acetone and HCL, methylene chloride and MEOH, DMSO and methanol. Solvent combinations for SPD140 P2 KIT are DMSO, DMF and toluene.
SPD140 P1 and P2 kits are recommended for solid phase extractions, drying lipid extractions, flash chromatography fractions, solid phase synthesis cleavage solutions such as protein hydrolyzates and evapourating synthesis solids, compound management, evapourating dissolution solvents.
Bestellinformation: SPD140P2 includes acid-resistant SpeedVac™ vacuum concentrator, universal vacuum source (–50 °C ), dram vials 24×1 (15×45 mm) 4 ml - dram vial rotor (RH24-15), chemical trap, tubing and fitting kit, disposable cartridge, and glass condensation flask.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge