Savant™ SpeedVac™ SPD300DDAA Large Capacity Vacuum Concentrator Kit
Lieferant: THERMO SAVANT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SPD300DDAA-230
THSASPD300DDAA-230EA 42000 EUR InStock
THSASPD300DDAA-230
Savant™ SpeedVac™ SPD300DDAA Large Capacity Vacuum Concentrator Kit
Konzentratoren
Thermo Scientific™ Savant™ SpeedVac™ SPD300DDA vacuum concentrator is a large-capacity vacuum concentrator with a modular design, ideal for drying aggressive organic solvents, strong acids, bases, and combinatorial chemistry solvents. It is capable of drying larger quantities of samples at one time and can support high-volume sample preparation needs.
  • Proven reliability: Backed by over 50 years of experience in the Savant SpeedVac family
  • Consistent Drying: Features four IR heat lamps, PTFE-coated chamber, and heated glass lid for efficient drying of various solvents and acids. Digital vacuum gauge prevents uncontrolled boiling
  • Convenience and flexibility: Universal rotor supports large-sample processing and multiple formats like microcentrifuge tubes and microplates
  • User-friendly: Interface with four preset and eight user-defined programs for optimal operation settings
  • Safety assurance: Secure glass lid, hands-free rotor loading, and audible alarm for enhanced safety
  • Sample integrity: Ability to download live run data from a USB port enhances data integrity
The SPD300DDAA-230 is a versatile surge protection device designed for critical laboratory and industrial applications, ensuring reliable operation and safeguarding equipment in processes such as solid-phase extractions, drying lipid extracts, flash chromatography fractions, solid-phase synthesis cleavage solutions, and high-performance liquid chromatography (HPLC). By protecting systems from transient overvoltages, the SPD300DDAA-230 maintains the integrity and efficiency of these essential processes, making it an invaluable asset for enhancing the quality and reliability of scientific and industrial outputs.
Bestellinformation: SPD300DDAA-230 includes Acid-resistant SpeedVac™ vacuum concentrator, Single-shelf univeral flask, 4-place flask/plate rotor accessory, Glass condensation flask, CryoCool™ heat transfer fluid (1 l).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge