Cation exchange chromatography columns, TSK-GEL® SP-2SW
Lieferant: TOSOH Bioscience
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0007165
TOYO0007165EA 2060 EUR InStock
TOYO0007165
Cation exchange chromatography columns, TSK-GEL® SP-2SW
Chromatographiesäulen
TSK-GEL® SP-2SW are silica based stainless steel column packed with spherical silica beads which are surface modified with sulfopropyl strong cation exchange groups. Designed for the separation and analysis of small protein, peptides, nucleosides and other biologically active molecules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge