Anion exchange chromatography columns, TSK-GEL®, BioAssist Q
Lieferant: TOSOH Bioscience
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0019685
TOYO0019685EA 1870 EUR InStock
TOYO0019685
Anion exchange chromatography columns, TSK-GEL®, BioAssist Q
Chromatographiesäulen
TSK-Gel® BioAssist Q are polymer based packed PEEK columns for laboratory applications. These columns provide high capacity for small to very large MW proteins and nucleic acids.
  • High capacity even for large proteins
  • Unique pore structure
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge