0007476
0007477
0007478
0014477
0014478
0014479
0014701
0014718
0014720
0016050
0016052
0016172
0016173
0019026
0043151
0043152
0043153
0043176
0043177
0043276
0043277
0019818
0044465
0014703
0019956
0021448
0021888
0043126
0019027
0019955
0019957
0016174
0014783
0014784
0014785
0016175
0043127
0021449
0021450
0021889
0021887
0019802
0021301
TOYO0007476EA
709
EUR
InStock
TOYO0007476
TOYO0007477
TOYO0007478
TOYO0014477
TOYO0014478
TOYO0014479
TOYO0014701
TOYO0014718
TOYO0014720
TOYO0016050
TOYO0016052
TOYO0016172
TOYO0016173
TOYO0019026
TOYO0043151
TOYO0043152
TOYO0043153
TOYO0043176
TOYO0043177
TOYO0043276
TOYO0043277
TOYO0019818
TOYO0044465
TOYO0014703
TOYO0019956
TOYO0021448
TOYO0021888
TOYO0043126
TOYO0019027
TOYO0019955
TOYO0019957
TOYO0016174
TOYO0014783
TOYO0014784
TOYO0014785
TOYO0016175
TOYO0043127
TOYO0021449
TOYO0021450
TOYO0021889
TOYO0021887
TOYO0019802
TOYO0021301
Hydrophobic interaction chromatography resins, Toyopearl®, TSK-GEL®
Chromatographie-Sorbentien
HPLC-Sorbentien
Hydrophobic interaction chromatography (HIC) is a powerful tool for the process purification of biomolecules. Five HIC ligands featuring different degrees of hydrophobicity and selectivity. The hydrophobicity of HIC resins increases through the ligand series.
- Typically used in intermediate purification and polishing steps
Ether HIC resins: Ether hydrophobic interaction chromatography (HIC) resins have the highest hydrophilicity (lowest hydrophobicity) of the HIC ligands. The ether ligand is useful for very hydrophobic molecules such as membrane proteins. It is typically used in intermediate purification and polishing steps.
Phenyl HIC resins: Phenyl hydrophobic interaction chromatography resins are in the middle of the hydrophobicity range. Primary applications are for the separation of proteins, their isoforms, and aggregate removal.
Butyl HIC resins: Butyl hydrophobic interaction chromatography (HIC) resins have the second highest hydrophobicity of the HIC ligands. There are three pore sizes available for this ligand and each has been optimised for a specific molecular size profile. The primary applications are the separation of proteins, their isoforms, and aggregate removal.
Hexyl HIC resins: Hexyl hydrophobic interaction chromatography (HIC) resin has the highest hydrophobicity of the HIC ligands. It is used for very hydrophilic proteins or where a low salt elution environment is required.