Cation exchange chromatography columns, TSK-GEL® CM STAT
Lieferant: TOSOH Bioscience
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0021965 0021966
TOYO0021965EA 1320 EUR InStock
TOYO0021965 TOYO0021966
Cation exchange chromatography columns, TSK-GEL® CM STAT
Chromatographiesäulen
TSK-GEL® CM STAT are cross-linked hydrophilic polymer (mono-disperse particles) based columns designed to analyse the profile of charge isomers of proteins. These STAT columns provide fast, high resolution separations at moderate pressures.
  • Nonporous columns
  • High efficiency separation at short analysis time
  • Used for high resolution protein separation
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge