Process development columns, ToyoScreen® RoboColumn®
Lieferant: TOSOH Bioscience
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0045001 0045002 0045003 0045004 0045005 0045006 0045007 0045008 0045021 0045022 0045023 0045024 0045025 0045026 0045031 0045032 0045033 0045034 0045035 0045036 0045037 0045038 0045051 0045052 0045061 0045062 0045063 0045064 0045071 0045072 0045027 0045028
TOYO0045001EA 356 EUR InStock
TOYO0045001 TOYO0045002 TOYO0045003 TOYO0045004 TOYO0045005 TOYO0045006 TOYO0045007 TOYO0045008 TOYO0045021 TOYO0045022 TOYO0045023 TOYO0045024 TOYO0045025 TOYO0045026 TOYO0045031 TOYO0045032 TOYO0045033 TOYO0045034 TOYO0045035 TOYO0045036 TOYO0045037 TOYO0045038 TOYO0045051 TOYO0045052 TOYO0045061 TOYO0045062 TOYO0045063 TOYO0045064 TOYO0045071 TOYO0045072 TOYO0045027 TOYO0045028
Process development columns, ToyoScreen® RoboColumn®
Chromatographiesäulen
Chromatographic columns pre-packed with TOYOPEARL ion exchange, mixed-mode, hydrophobic interaction or affinity media.
  • High throughput parallel chromatography
  • Automated screening of resins and parameters
  • Liquid flow in the columns is driven by positive pressure liquid displacement
These columns can be operated with a robotic liquid handling system, such as the Freedom EVO® from TECAN. This allows automated high-throughput, small-scale bio chromatographic separations of protein samples by running up to eight individual columns simultaneously.
Lieferumfang: Columns are supplied in a row of eight units pre-packed with the TOYOPEARL resin and sealed with two removable silicon cover seals for proper storage.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge