Secure cabinets for pesticides product
Lieferant: TRIONYX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AL302
TRIOAL302EA 663 EUR InStock
TRIOAL302
Secure cabinets for pesticides product
Möbel Lagerschränke
The cabinets is recommended for storage of herbicides, fungicides, insecticides and other phytosanitary products. Also recommended for farmers, ranchers, growers, landscapers, growers, agricultural cooperatives, green spaces, community etc. It allows a safety storage of phytosanitary products.
  • Hight and low vents
  • Double door with two key locking points
Zertifizierungen: Large warning labels in compliance with ISO 3864 norm and 92/58/CEE european directive.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge