Tray
Lieferant: TECHNICAL QUARTZ SOLUTIONS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
73034
TSLA73034EA 279 EUR InStock
TSLA73034
Tray
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Trays made of quartz.
  • Temperature resistance up to +1050 °C
  • Highly resistant to chemicals
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge