Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Evaporating Dishes with Spout

Lieferant: TECHNICAL QUARTZ SOLUTIONS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

75000 75001 75003 75004 75006 75008 75010 75015
TSLA75000EA 32.4 EUR InStock
TSLA75000 TSLA75001 TSLA75003 TSLA75004 TSLA75006 TSLA75008 TSLA75010 TSLA75015
Evaporating Dishes with Spout
Schalen/Schüsseln/Töpfe Abdampfschalen
Temperature resistant evaporating dishes made from transparent quartz glass.

  • Excellent chemical resistance
  • High temperature resistance
  • Made for tasks up to 1050 °C
  • High purity

The TQS quartz glass evaporating dishes are made for tasks up to 1050 °C (maximum limit 1280 °C). High chemical resistance and purity characterize these products. Quartz glass products have a long life for frequent use with temperatures up to 1050 °C but the durabilty is dependent on its use and application.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge