Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Crystallising Dishes, with Spout

Lieferant: TECHNICAL QUARTZ SOLUTIONS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

75150 75151 75152 75153 75154 75155
TSLA75150EA 37.2 EUR InStock
TSLA75150 TSLA75151 TSLA75152 TSLA75153 TSLA75154 TSLA75155
Crystallising Dishes, with Spout
Schalen/Schüsseln/Töpfe Kristallisierschalen
Temperature-resistant crystallising dishes made from transparent quartz glass.

  • Excellent chemical resistance
  • High resistance against thermal shock
  • Made for tasks up to 1050 °C
  • High purity

The TQS quartz glass crystallising dishes are made for tasks up to 1050 °C (maximum limit 1280 °C). High chemical resistance and purity characterise these products. Quartz glass products have a long life for frequent use with temperatures up to 1050 °C, but the durability is dependant upon use and applications.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge