Sample loops for high pressure valves, stainless steel, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1876
UPCH1876EA 532 EUR InStock
UPCH1876
Sample loops for high pressure valves, stainless steel, Upchurch Scientific®
Chromatographie-Zubehör Chromatographie-Probenschläuche
These high quality stainless steel sample loops have burr-free, square-cut ends to ensure a flush connection to valve ports.
  • Size designations of loops are nominal
  • Supplied with fittings that are not swaged onto the tube
The loop has to be completely bottomed in the injector port before the ferrule is swaged onto the tube.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge